Learn More
Sigma Aldrich Fine Chemicals Biosciences tobaco etch virus TEV Proteas
Catalog No. 502900935
Encompass
The tobacco etch virus (TEV) protease is a useful tool for the removal of fusion tags from recombinant proteins.
Catalog No. 50-290-0935 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T44551KU
The tobacco etch virus (TEV) protease is a useful tool for the removal of fusion tags from recombinant proteins.