Learn More
Enzo Life Sciences Tobramycin (1g). CAS: 32986-56-4
SDP
Catalog No. 502008911
Encompass_Preferred
Aminoglycoside antibiotic with similar structure to the kanamycins and the gentamycins. Used in the treatment of respiratory infection and cystic fibrosis. Alternative name: Deoxykanamycin B, Distobram, Gernebcin, Obramycin, Tobradistin. Formulation: Free base. Appearance: White to off-white powder. Solubility: Soluble in water; very slightly soluble in 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-8911 Supplier Enzo Life Sciences Supplier No. ALX380018G001
Aminoglycoside antibiotic with similar structure to the kanamycins and the gentamycins. Used in the treatment of respiratory infection and cystic fibrosis. Alternative name: Deoxykanamycin B, Distobram, Gernebcin, Obramycin, Tobradistin. Formulation: Free base. Appearance: White to off-white powder. Solubility: Soluble in water; very slightly soluble in 100% ethanol. Long Term Storage: +4°C.