Learn More
Enzo Life Sciences Tobramycin (250mg). CAS: 32986-56-4
SDP
Supplier:  Enzo Life Sciences ALX380018M250
Encompass_Preferred
Aminoglycoside antibiotic with similar structure to the kanamycins and the gentamycins. Used in the treatment of respiratory infection and cystic fibrosis. Alternative name: Deoxykanamycin B, Distobram, Gernebcin, Obramycin, Tobradistin. Formulation: Free base. Solubility: Soluble in water; very slightly soluble in 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-8912