Learn More
Medchemexpress LLC Tobramycin | 32986-56-4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04415G
Encompass_Preferred
Tobramycin (Nebramycin Factor 6) is a broad-spectrum aminoglycoside antibiotic commonly used to treat moderate to severe bacterial infections caused by sensitive organisms. It is also utilized in pneumonia research, particularly for infections involving Pseudomonas aeruginosa, and can decrease P. aeruginosa cell viability.
  • Broad-spectrum aminoglycoside antibiotic
  • Administered parenterally
  • Treats moderate to severe bacterial infections
  • Effective in pneumonia research, especially against Pseudomonas aeruginosa
  • Decreases P. aeruginosa cell viability
  • Demonstrates bacterial cell kill effects in animal models
Catalog No. 50-004-08225