Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tobramycin | 32986-56-4 | 99.9% | 10 G
SDP

Catalog No. 5000408771
Encompass_Preferred

Tobramycin (Nebramycin Factor 6) is a broad-spectrum aminoglycoside antibiotic, parenterally administered for moderate to severe bacterial infections due to sensitive organisms. It is also used in pneumonia research related to Pseudomonas aeruginosa.

  • Target: Bacterial; antibiotic
  • Pathway: Anti-infection
  • Molecular formula: C18H37N5O9
  • Molecular weight: 467.51
  • Solubility in H2O: ≥ 100 mg/mL
  • Solubility in DMSO: 2 mg/mL (needs ultrasonic)
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-08771 Supplier Medchemexpress LLC Supplier No. HYB044110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tobramycin (Nebramycin Factor 6) is a broad-spectrum aminoglycoside antibiotic, parenterally administered for moderate to severe bacterial infections due to sensitive organisms. It is also used in pneumonia research related to Pseudomonas aeruginosa.

  • Target: Bacterial; antibiotic
  • Pathway: Anti-infection
  • Molecular formula: C18H37N5O9
  • Molecular weight: 467.51
  • Solubility in H2O: ≥ 100 mg/mL
  • Solubility in DMSO: 2 mg/mL (needs ultrasonic)
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.