Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tobramycin | 32986-56-4 | 99.9% | 25 G
SDP

Catalog No. 5000405626
Encompass_Preferred

Tobramycin is a broad-spectrum aminoglycoside antibiotic used to treat moderate to severe bacterial infections caused by sensitive organisms. It is also used in pneumonia research, particularly related to *Pseudomonas aeruginosa*.

  • Parenterally administered antibiotic
  • Treats moderate to severe bacterial infections
  • Used in pneumonia research related to *Pseudomonas aeruginosa*
  • Purity of 99.9%
  • Available as a solid
  • Soluble in water and DMSO

Catalog No. 50-004-05626 Supplier Medchemexpress LLC Supplier No. 25GHYB0441
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tobramycin is a broad-spectrum aminoglycoside antibiotic used to treat moderate to severe bacterial infections caused by sensitive organisms. It is also used in pneumonia research, particularly related to *Pseudomonas aeruginosa*.

  • Parenterally administered antibiotic
  • Treats moderate to severe bacterial infections
  • Used in pneumonia research related to *Pseudomonas aeruginosa*
  • Purity of 99.9%
  • Available as a solid
  • Soluble in water and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.