Learn More
Medchemexpress LLC Tobramycin | 32986-56-4 | 99.9% | 500 MG
SDP
Catalog No. 5000408228
Encompass_Preferred
Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad-spectrum aminoglycoside antibiotic used to treat moderate to severe bacterial infections caused by sensitive organisms. It is also used in pneumonia research, particularly for infections caused by Pseudomonas aeruginosa.
  • Aminoglycoside antibiotic
  • Broad-spectrum activity
  • Used for bacterial infections
  • Useful in pneumonia research for Pseudomonas aeruginosa
  • Decreases P. aeruginosa cell viability when combined with mycobacterium fortuitum enzyme (PodA)
  • Produces bacterial cell kill effect in murine pneumonia models when combined with Meropenem
Catalog No. 50-004-08228 Supplier Medchemexpress LLC Supplier No. HYB0441500MG
Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad-spectrum aminoglycoside antibiotic used to treat moderate to severe bacterial infections caused by sensitive organisms. It is also used in pneumonia research, particularly for infections caused by Pseudomonas aeruginosa.
  • Aminoglycoside antibiotic
  • Broad-spectrum activity
  • Used for bacterial infections
  • Useful in pneumonia research for Pseudomonas aeruginosa
  • Decreases P. aeruginosa cell viability when combined with mycobacterium fortuitum enzyme (PodA)
  • Produces bacterial cell kill effect in murine pneumonia models when combined with Meropenem