Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tobramycin | 32986-56-4 | MFCD00077885 | 99.9% | 100 MG
SDP

Catalog No. 5000407055
Encompass_Preferred

Tobramycin is a parenterally administered, broad-spectrum aminoglycoside antibiotic. It is widely used in the treatment of moderate to severe bacterial infections caused by sensitive organisms. It can also be used for pneumonia research caused by *Pseudomonas aeruginosa*.

  • Broad-spectrum aminoglycoside antibiotic
  • Treats bacterial infections and is used in pneumonia research
  • Soluble in water (≥ 100 mg/mL), DMSO (2 mg/mL), and PBS (100 mg/mL)
  • Targets bacterial organisms as an antibiotic
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-07055 Supplier Medchemexpress LLC Supplier No. HYB0441100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tobramycin is a parenterally administered, broad-spectrum aminoglycoside antibiotic. It is widely used in the treatment of moderate to severe bacterial infections caused by sensitive organisms. It can also be used for pneumonia research caused by *Pseudomonas aeruginosa*.

  • Broad-spectrum aminoglycoside antibiotic
  • Treats bacterial infections and is used in pneumonia research
  • Soluble in water (≥ 100 mg/mL), DMSO (2 mg/mL), and PBS (100 mg/mL)
  • Targets bacterial organisms as an antibiotic
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.