Learn More
Research Products International Corp Tobramycin 5 G
SDP
Catalog No. 502230548
Encompass_Preferred
Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections. It is effective against a range of gram-negative bacteria.
Tobramycin works by inhibiting bacterial protein synthesis. It binds to the bacterial ribosomes, disrupting the synthesis of proteins essential for bacterial growth and survival.
It is particularly effective against Gram-negative bacteria, including Pseudomonas aeruginosa, Escherichia coli, Klebsiella pneumoniae, and others. It is less effective against Gram-positive bacteria.
Catalog No. 50-223-0548 Supplier Research Products International Corp Supplier No. T521505.0
Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections. It is effective against a range of gram-negative bacteria.
Tobramycin works by inhibiting bacterial protein synthesis. It binds to the bacterial ribosomes, disrupting the synthesis of proteins essential for bacterial growth and survival.
It is particularly effective against Gram-negative bacteria, including Pseudomonas aeruginosa, Escherichia coli, Klebsiella pneumoniae, and others. It is less effective against Gram-positive bacteria.