Learn More
Selleck Chemical LLC Tobramycin S2514-50mg
SDP
Catalog No. 503110114
Encompass_Preferred
Tobramycin (NSC 180514 Deoxykanamycin B Nebramycin Factor 6) is an aminoglycoside antibiotic with an IC50 of 9 7 M
Catalog No. 50-311-0114 Supplier Selleck Chemical LLC Supplier No. S251450MG
Tobramycin (NSC 180514 Deoxykanamycin B Nebramycin Factor 6) is an aminoglycoside antibiotic with an IC50 of 9 7 M