Learn More
Abcam Tobramycin, free base, Ribosomal translation inhibitor, 500MG
Catalog No. 502707377
Encompass_Preferred
MW 467.5 Da, Purity >98%. Inhibits ribosomal translation. Aminoglycoside antibiotic which binds 30S ribosomal subunit A, preventing functional protein production. Effective against gram-negative bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7377 Supplier Abcam Supplier No. AB120659500MG
MW 467.5 Da, Purity >98%. Inhibits ribosomal translation. Aminoglycoside antibiotic which binds 30S ribosomal subunit A, preventing functional protein production. Effective against gram-negative bacteria.
The product is subject to the following: Abcam Restricted Use Statement