Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin, MP Biomedicals™

Catalog No. MP215692691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215692691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215692691 Supplier MP Biomedicals, Inc Supplier No. 0215692691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Effective against gram negative bacteria, especially the Pseudomonas species. Tobramycin, MP Biomedicals is an aminoglycoside antibiotic that binds to the 70S ribosomal subunit. It inhibits translocation and hence elicits miscoding.

Tobramycin is an aminoglycoside, broad-spectrum antibiotic derived from Streptomyces tenebrarius. It is used to treat a variety of bacterial infections. Tobramycin is preferred over gentamicin for Pseudomonas aeruginosa pneumonia due to better lung penetration. It is not effective against Enterococci.

It is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis, and other soft-tissue infections. It is a potential therapy for sinus infections.

Tobramycin inhibits bacterial protein synthesis. It works by binding to a site on bacterial 30S and 50S ribosome, preventing formation of the 70S complex. As a result, mRNA cannot be translated into protein and cell death ensues.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1?6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1?4)]-2-deoxy-D-streptamine, Deoxykanamycin B , Gotabiotic , Nebramycin
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water (50mg/mL).
Formula Weight 467.51
Grade Not Available
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.