Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin, MP Biomedicals™

Catalog No. MP219969790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219969790 500 mg
MP219969780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219969790 Supplier MP Biomedicals, Inc Supplier No. 0219969790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tobramycin USP, Off-white Powder

  • Tobramycin has been used:
  • it is used to study the effect of tobramycin on bacteriophage infected biofilms
  • it is used to study the effects of antibiotics on Pseudomonas aeruginosa strain
  • it is used as a comparator antibiotic. Tobramycin is an aminoglycoside, broad-spectrum antibiotic produced by Streptomyces tenebrarius.
  • It is effective against gram-negative bacteria, especially the Pseudomonas species.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
MDL Number MFCD00077885
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine

Specifications

Melting Point 178°C
Color Off-white
pH 9 to 11 (10%)
Quantity 500 mg
Beilstein 1357507
Solubility Information H2O: soluble (50mg/mL)
Physical Form Powder
storageNote1 +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.