Learn More
Alkali Scientific Tobramycin [O 3 Amino 3 deoxy α D glucopyranosyl (1 6) O [2, 6 diamino 2, 3, 6 trideoxy α D ribohexopyranosyl (1 4)] 2 deoxy D streptamine], 5 Grams
SDP
Catalog No. 5000661022
Encompass_Preferred
Tobramycin is an aminoglycoside antibiotic used to treat a variety of bacterial infections. This 5g powder form is commonly used in laboratories for research purposes, particularly in microbiological assays testing its antibacterial activity. Tobramycin inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, thereby preventing the bacterial cell from producing essential proteins required for growth. It is primarily effective against Gram-negative bacteria, including Pseudomonas aeruginosa and other resistant organisms. This research-grade tobramycin is widely utilized in studies examining microbial resistance mechanisms and in vitro testing of its therapeutic potential.
Catalog No. 50-006-61022 Supplier Alkali Scientific Supplier No. TOB001CP
Tobramycin is an aminoglycoside antibiotic used to treat a variety of bacterial infections. This 5g powder form is commonly used in laboratories for research purposes, particularly in microbiological assays testing its antibacterial activity. Tobramycin inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, thereby preventing the bacterial cell from producing essential proteins required for growth. It is primarily effective against Gram-negative bacteria, including Pseudomonas aeruginosa and other resistant organisms. This research-grade tobramycin is widely utilized in studies examining microbial resistance mechanisms and in vitro testing of its therapeutic potential.