Learn More
Alkali Scientific Tobramycin Sulfate, 1 G
SDP
Catalog No. 5000661039
Encompass_Preferred
Tobramycin Sulfate, 1 Gram, is a potent antibiotic used for the treatment of serious infections caused by gram-negative bacteria, including Pseudomonas aeruginosa. Available in powder form, it is typically reconstituted and administered intravenously in clinical settings. The 1-gram dosage is suitable for severe infections, ensuring effective treatment for high-risk patients. Tobramycin works by inhibiting bacterial protein synthesis, preventing bacterial growth and replication. It is often used for infections in immunocompromised patients, in burn units, and for patients with cystic fibrosis, as well as in other life-threatening bacterial infections.
Catalog No. 50-006-61039 Supplier Alkali Scientific Supplier No. TOB003CP
Tobramycin Sulfate, 1 Gram, is a potent antibiotic used for the treatment of serious infections caused by gram-negative bacteria, including Pseudomonas aeruginosa. Available in powder form, it is typically reconstituted and administered intravenously in clinical settings. The 1-gram dosage is suitable for severe infections, ensuring effective treatment for high-risk patients. Tobramycin works by inhibiting bacterial protein synthesis, preventing bacterial growth and replication. It is often used for infections in immunocompromised patients, in burn units, and for patients with cystic fibrosis, as well as in other life-threatening bacterial infections.