Learn More
Alkali Scientific Tobramycin Sulfate, 100 Mg
SDP
Catalog No. 5000660510
Encompass_Preferred
Tobramycin Sulfate is an aminoglycoside antibiotic used for treating severe bacterial infections caused by gram-negative bacteria. Available in powder form, it is commonly used for the treatment of respiratory tract infections, urinary tract infections, and other life-threatening bacterial infections. The 100mg vial provides a concentrated dose to ensure the antibiotic's efficacy against resistant organisms. Tobramycin works by inhibiting bacterial protein synthesis, making it effective for treating infections that do not respond to other antibiotics. The product is commonly used in hospitals and clinical settings for intravenous administration.
Catalog No. 50-006-60510 Supplier Alkali Scientific Supplier No. TOB002CP
Tobramycin Sulfate is an aminoglycoside antibiotic used for treating severe bacterial infections caused by gram-negative bacteria. Available in powder form, it is commonly used for the treatment of respiratory tract infections, urinary tract infections, and other life-threatening bacterial infections. The 100mg vial provides a concentrated dose to ensure the antibiotic's efficacy against resistant organisms. Tobramycin works by inhibiting bacterial protein synthesis, making it effective for treating infections that do not respond to other antibiotics. The product is commonly used in hospitals and clinical settings for intravenous administration.