Learn More
Research Products International Corp TOBRAMYCIN SULFATE, 100MG
SDP
Catalog No. 50213696
Encompass_Preferred
Tobramycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, leading to the misreading of mRNA and preventing the proper initiation complex formation. This interference disrupts the translation process and inhibits bacterial growth.
Tobramycin is used in microbiology for selective bacterial isolation and in antibiotic sensitivity testing to determine the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.
WARNING! This product can expose you to Tobramycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-696 Supplier Research Products International Corp Supplier No. T450000.1
Tobramycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, leading to the misreading of mRNA and preventing the proper initiation complex formation. This interference disrupts the translation process and inhibits bacterial growth.
Tobramycin is used in microbiology for selective bacterial isolation and in antibiotic sensitivity testing to determine the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.
WARNING! This product can expose you to Tobramycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.