Learn More
Medchemexpress LLC Tobramycin sulfate | 49842-07-1 | 98.0% | 500 MG
SDP
Catalog No. 5000406051
Encompass_Preferred
Tobramycin sulfate is a broad-spectrum aminoglycoside antibiotic administered parenterally. It is commonly used in the treatment of moderate to severe bacterial infections caused by sensitive organisms and can be utilized in pneumonia research specifically related to Pseudomonas aeruginosa.
  • Targets aminoglycoside
  • Decreases P. aeruginosa cell viability in combination with Mycobacterium fortuitum enzyme
  • Produces bacterial cell kill effect in murine pneumonia models
Catalog No. 50-004-06051 Supplier Medchemexpress LLC Supplier No. HYB0441A500MG
Tobramycin sulfate is a broad-spectrum aminoglycoside antibiotic administered parenterally. It is commonly used in the treatment of moderate to severe bacterial infections caused by sensitive organisms and can be utilized in pneumonia research specifically related to Pseudomonas aeruginosa.
  • Targets aminoglycoside
  • Decreases P. aeruginosa cell viability in combination with Mycobacterium fortuitum enzyme
  • Produces bacterial cell kill effect in murine pneumonia models