Learn More
Alkali Scientific Tobramycin Sulfate, 5 G
SDP
Catalog No. 5000661040
Encompass_Preferred
Tobramycin Sulfate 5 Gram is a high-dose antibiotic formulation used for treating severe infections caused by resistant gram-negative bacteria. The powder form is ideal for reconstitution and intravenous administration in hospitalized patients. The 5-gram vial is typically used for treating systemic infections such as pneumonia, septicemia, and urinary tract infections. As an aminoglycoside, Tobramycin inhibits bacterial protein synthesis, preventing bacterial replication. This dosage is suitable for critically ill patients requiring high antibiotic concentrations to combat multi-drug resistant organisms. The product is used in intensive care and surgical settings.
Catalog No. 50-006-61040 Supplier Alkali Scientific Supplier No. TOB004CP
Tobramycin Sulfate 5 Gram is a high-dose antibiotic formulation used for treating severe infections caused by resistant gram-negative bacteria. The powder form is ideal for reconstitution and intravenous administration in hospitalized patients. The 5-gram vial is typically used for treating systemic infections such as pneumonia, septicemia, and urinary tract infections. As an aminoglycoside, Tobramycin inhibits bacterial protein synthesis, preventing bacterial replication. This dosage is suitable for critically ill patients requiring high antibiotic concentrations to combat multi-drug resistant organisms. The product is used in intensive care and surgical settings.