Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin Sulfate, G-Biosciences
SCI_ED

Catalog No. S157221
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157221 1 g
S139086 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157221 Supplier G-Biosciences Supplier No. RC1121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tobramycin is an aminoglycoside antibiotic isolated from Streptomyces tenebrarius targeting gram-negative bacteria. It is often used to combat Pseudomonas species

It has been used to study cystic fibrosis and a variety of respiratory infections.

Chemical Identifiers

CAS 79645-27-5 , 49842-07-1

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 1 g
Percent Purity 98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.