Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin Sulfate USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048136 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
1 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-136 25 g
18-048-056 1 g
18-048-099 100 g
18-048-100 500 g
18-048-109 5 g
18-048-116 1 kg
18-048-135 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-048-136 Supplier Medisca Supplier No. 031904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79645-27-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White to Light Yellow
Quantity 25 g
Percent Purity 634 to 739 μg of Tobramycin/mg
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.