Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin Sulfate, USP, Spectrum™ Chemical
SDP

Catalog No. 18607431 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-431 5 g Amber Glass Bottle
18-607-432 25 g Amber Glass Bottle
18-607-433 50 g Amber Glass Bottle
18-607-430 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-431 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T15985GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

T1598, 79645-27-5, (C18H37N5O9)2·5H2SO4

Spectrum™ Chemical Tobramycin Sulfate, USP belongs to a class of drugs known as aminoglycoside antibiotics that works by stopping the growth of bacteria. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 79645-27-5
Molecular Formula C18H39N5O13S
Molecular Weight (g/mol) 565.59
MDL Number MFCD00133864,MFCD00133864
InChI Key ZEUUPKVZFKBXPW-UHFFFAOYNA-N
IUPAC Name 4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
SMILES OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O

Specifications

Physical Form Crystalline Powder
pH 6.0 to 8.0
CAS Max % 1
Ignition Residue 0.01
Packaging Amber Glass Bottle
Water 0.02
Color White or White/Yellow
Quantity 5 g
Percent Purity 97 to 100%
Grade USP
Chemical Name or Material Triclosan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.