Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company TOC 5.6MM OD X 2.4MM ID 1METER

Catalog No. NC2258829 Shop All Hach Company Products
Encompass

TOC ANALYZER TUBING, EMPP TUBING 5.6MM OD X 2.4MM ID, LENGTH: 1 METER

This item is not returnable. View return policy
Catalog No. NC2258829 Supplier Hach Company Supplier No. 10REH003
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TOC ANALYZER TUBING, EMPP TUBING 5.6MM OD X 2.4MM ID, LENGTH: 1 METER

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.