Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions TOC Standard 8000 ppm | MFCD02100762 | 4 L
SDP

Catalog No. NC1492700
Encompass_Preferred

TOC Standard 8000 ppm | MFCD02100762 | 4 L

Catalog No. NC1492700 Supplier Aqua Solutions Supplier No. 95884L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TOC Standard 8000 ppm | MFCD02100762 | 4 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.