Learn More
VEOLIA WTS TOC ACCURACY/PREC / VER SET
Catalog No. NC2894282
Encompass
M500/500/860 Acc/Prec Ver Set TOC OnlyAccuracy/Precision Verification set (TOC only) forSievers M500 and 500 RL TOC Analyzers with Standard iOS and Sievers TOC860 Analyzer. Includes one Verification Blank and one 500 µg C/L TOCVerification Standard (as NIST Sucrose). Conductivity standards are notincluded in this set.1 EA
Catalog No. NC2894282 Supplier VEOLIA WTS Supplier No. STD 74407
M500/500/860 Acc/Prec Ver Set TOC OnlyAccuracy/Precision Verification set (TOC only) forSievers M500 and 500 RL TOC Analyzers with Standard iOS and Sievers TOC860 Analyzer. Includes one Verification Blank and one 500 µg C/L TOCVerification Standard (as NIST Sucrose). Conductivity standards are notincluded in this set.1 EA