Learn More
VEOLIA WTS TOC CALIBRATION SET A
Supplier:  VEOLIA WTS STD 74405
Encompass
M500/500/860 Single Pt Cal Set A TOC OnlySingle point calibration set A (TOC only) for SieversM500 and 500 RL TOC Analyzers with Standard iOS and Sievers 860 TOCAnalyzer. Includes two Calibration Blanks and one 1.5 mg C/L TOC CalibrationStandard (as KHP). Conductivity standards are not included in this set.1 EA
Catalog No. NC2894280