Learn More
VEOLIA WTS TOC CALIBRATION SET B
Supplier:  VEOLIA WTS STD 74404
Encompass
M500/500 Multi-Point Cal Set BNOT TO BE SOLD TO CUSTOMERS. INTERNAL USEONLY. Multi-point calibration set B for Sievers M500 and 500 series analyzers withStandard iOS. Includes one Calibration Blank and two TOC CalibrationStandards (500 µg C/L and 1.0 mg C/L as KHP)
Catalog No. NC2894281