Learn More
Absolute Standards TOC Calibration Standard ©, 877-24-7, 1000 µg/mL
SDP
Catalog No. NC9955790
Encompass
TOC Calibration Standard ©, 877-24-7, 1000 µg/mL, Solvent in Water
Catalog No. NC9955790 Supplier Absolute Standards Supplier No. 59179
TOC Calibration Standard ©, 877-24-7, 1000 µg/mL, Solvent in Water