Learn More
Parker Hannifan Parker Balston TOC-1250 TOC Gas Generator, High-Purity Gas Supply, Cost-Effective Alternative, 1 Unit
SDP
Catalog No. NC1980121
Encompass
The Parker Balston TOC-1250 is a benchtop TOC Gas Generator that produces high-purity, hydrocarbon-free, and CO2-free carrier gas for TOC analyzers. It eliminates reliance on high-pressure gas cylinders, ensuring consistent and reliable analysis with low operating costs and minimal environmental impact. This system uses advanced purification technologies, making it a safer and economical choice compared to traditional gas supplies.
Catalog No. NC1980121 Supplier Parker Hannifan Supplier No. TOC1250
Add to Cart
Add to Cart
Request a Quote
The Parker Balston TOC-1250 is a benchtop TOC Gas Generator that produces high-purity, hydrocarbon-free, and CO2-free carrier gas for TOC analyzers. It eliminates reliance on high-pressure gas cylinders, ensuring consistent and reliable analysis with low operating costs and minimal environmental impact. This system uses advanced purification technologies, making it a safer and economical choice compared to traditional gas supplies.