Learn More
Hach Company Total Organic Carbon (TOC) Reagent Set, HR
Supplier:  Hach Company 2760445
Encompass
For determination of Total Organic Carbon (TOC) by the High Range Test 'N Tube™ method. Hach Method 10128. Range:100 - 700 mg/L C Set includes digestion vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed). Requires digestion.
Catalog No. 50-199-6640