Learn More
Hach Company Total Organic Carbon (TOC) Reagent Set, LR
Supplier:  Hach Company 2760345
Encompass
For determination of Total Organic Carbon (TOC) by the Low Range Test 'N Tube™ method. Hach Method 10129. Range: 0.3 - 20.0 mg/L C Set includes digestion vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed). Requires digestion.
Catalog No. 50-199-6639