Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions TOC Standard 100 ppm | MFCD02100762 | 100 mL
SDP

Catalog No. NC0951582
Encompass_Preferred

TOC Standard 100 ppm | MFCD02100762 | 100 mL

Catalog No. NC0951582 Supplier Aqua Solutions Supplier No. 9572100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TOC Standard 100 ppm | MFCD02100762 | 100 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.