Learn More
Aqua Solutions TOC STANDARD 1000 PPM 8 OZ
SDP
Catalog No. NC0173483
Encompass_Preferred
Total Organic Carbon Standard 1000 Ppm Solution, Prepared To Epa 415/ Standard Methods (250ml)
Catalog No. NC0173483 Supplier Aqua Solutions Supplier No. 9570250ML
Total Organic Carbon Standard 1000 Ppm Solution, Prepared To Epa 415/ Standard Methods (250ml)