Learn More
Aqua Solutions TOC STANDARD 200PPM 500 ML
SDP
Catalog No. NC9656600
Encompass_Preferred
Total Organic Carbon Standard 200 ppm Solution Prepared to EPA 415 / Standard Methods, 500 mL, Water Soluble, Appearance And Odors: Clear Odorless Liquid, Specific Gravity: 1, Volatile By Volume: >99%.
Catalog No. NC9656600 Supplier Aqua Solutions Supplier No. 9566500ML
Total Organic Carbon Standard 200 ppm Solution Prepared to EPA 415 / Standard Methods, 500 mL, Water Soluble, Appearance And Odors: Clear Odorless Liquid, Specific Gravity: 1, Volatile By Volume: >99%.