Learn More
Agilent Technologies Total organic carbon (TOC) standard, 500 mL
Catalog No. NC1756055
Encompass
Single component mixture, organic, water (low TOC, 50 ppb) solvent, and potassium hydrogen phthalate (as TOC): 500 mg/L concentration.
Catalog No. NC1756055 Supplier Agilent Technologies Supplier No. IQC1055
Single component mixture, organic, water (low TOC, 50 ppb) solvent, and potassium hydrogen phthalate (as TOC): 500 mg/L concentration.