Learn More
LaMarKa Chemical Company TOC Standard 1000ppm (preserved with Sulfuric)
Catalog No. NC9696853
Encompass_Preferred
Toc Standard 1000 Ppm Khp (Preserved With Sulfuric)-Bottle 16Oz Plastic
Catalog No. NC9696853 Supplier LaMarKa Chemical Company Supplier No. 00005823167482411
Toc Standard 1000 Ppm Khp (Preserved With Sulfuric)-Bottle 16Oz Plastic