Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Total Organic Carbon Std. 100 ppm Certified (EPA 415-1)(1L)
SDP

Catalog No. NC9692261
Encompass_Preferred

Total Organic Carbon Std. 100 ppm Certified (EPA 415-1)(1L)

Catalog No. NC9692261 Supplier Aqua Solutions Supplier No. 95721L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Total Organic Carbon Std. 100 ppm Certified (EPA 415-1)(1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.