Learn More
Hach Company TOC TNTplus Vial Test, HR (30-300 mg/L C), 25 Tests
Supplier:  Hach Company TNT811
Encompass
TOC TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Total Organic Carbon (TOC) by the Direct Method.
Catalog No. 50-199-8106