Learn More
Medchemexpress LLC Tocainide hydrochloride | 71395-14-7 | 99.5% | 1 ML
SDP
Catalog No. 5000412892
Encompass_Preferred
Tocainide hydrochloride is a sodium channel blocker that blocks sodium channels in pain-producing foci in nerve membranes. It is a primary amine analog of lidocaine and can be used for the treatment of tinnitus.
  • Suppresses ventricular ectopic activity.
  • Suitable for laboratory centrifugation and storage.
Catalog No. 50-004-12892 Supplier Medchemexpress LLC Supplier No. HYB1798A10MM/1ML
Tocainide hydrochloride is a sodium channel blocker that blocks sodium channels in pain-producing foci in nerve membranes. It is a primary amine analog of lidocaine and can be used for the treatment of tinnitus.
  • Suppresses ventricular ectopic activity.
  • Suitable for laboratory centrifugation and storage.