Learn More
Medchemexpress LLC Tocainide hydrochloride | 71395-14-7 | 99.5% | 5 MG
SDP
Catalog No. 5000413032
Encompass_Preferred
Tocainide hydrochloride is a sodium channel blocker that blocks sodium channels in the pain-producing foci in nerve membranes. It is a primary amine analog of lidocaine and can be used for the treatment of tinnitus.
  • Sodium channel blocker.
  • Blocks sodium channels in pain-producing foci in nerve membranes.
  • Can be used for the treatment of tinnitus.
Catalog No. 50-004-13032 Supplier Medchemexpress LLC Supplier No. HYB1798A5MG
Tocainide hydrochloride is a sodium channel blocker that blocks sodium channels in the pain-producing foci in nerve membranes. It is a primary amine analog of lidocaine and can be used for the treatment of tinnitus.
  • Sodium channel blocker.
  • Blocks sodium channels in pain-producing foci in nerve membranes.
  • Can be used for the treatment of tinnitus.