Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tocainide hydrochloride | 71395-14-7 | MFCD00941502 | 99.5% | 228.72 g/mol | C11H17ClN2O | 10 MG
SDP

Catalog No. 5000297244
Encompass_Preferred

Tocainide hydrochloride is a sodium channel blocker provided as a high-purity hydrochloride salt for research use. It is supplied as a solid off-white to gray powder and is intended for biochemical and pharmacological studies involving sodium channel modulation.

  • High purity suitable for research applications.
  • Sodium channel blocker used in electrophysiology and pharmacology studies.
  • Off-white to gray solid physical form.
  • Recommended storage: 4°C for solid; in solvent, -80°C for 6 months or -20°C for 1 month under nitrogen.
  • Molecular weight 228.72 g/mol and CAS number 71395-14-7 for identification.

Catalog No. 50-002-97244 Supplier Medchemexpress LLC Supplier No. HYB1798A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tocainide hydrochloride is a sodium channel blocker provided as a high-purity hydrochloride salt for research use. It is supplied as a solid off-white to gray powder and is intended for biochemical and pharmacological studies involving sodium channel modulation.

  • High purity suitable for research applications.
  • Sodium channel blocker used in electrophysiology and pharmacology studies.
  • Off-white to gray solid physical form.
  • Recommended storage: 4°C for solid; in solvent, -80°C for 6 months or -20°C for 1 month under nitrogen.
  • Molecular weight 228.72 g/mol and CAS number 71395-14-7 for identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.