Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tocainide hydrochloride, 98%

Catalog No. AAH3626503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36265-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH36265-03 Supplier Thermo Scientific™ Supplier No. H3626503
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71395-14-7
Molecular Formula C11H17ClN2O
Molecular Weight (g/mol) 228.72
MDL Number MFCD00941502
InChI Key AMZACPWEJDQXGW-UHFFFAOYNA-N
IUPAC Name 2-amino-N-(2,6-dimethylphenyl)propanamide hydrochloride
SMILES Cl.CC(N)C(=O)NC1=C(C)C=CC=C1C

Specifications

Quantity 1 g
Formula Weight 228.72
Percent Purity 98%
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 275-361-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.