Learn More
Medchemexpress LLC Toceranib-d8 | 1795134-78-9 | 99.1% | 404.51 g/mol | C22H17D8FN4O2 | 1 MG
SDP
Catalog No. 5000244644
Encompass_Preferred
Toceranib-d8 is the deuterium-labeled isotopologue of toceranib, provided as a research-grade analytical standard for laboratory use. It is intended for use as a stable isotope internal standard in LC-MS/MS, pharmacokinetic, and tracer studies to enable accurate quantitation and differentiation from the unlabeled compound.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (>99%) suitable for analytical applications.
  • Intended for pharmacokinetic, bioanalytical, and tracer studies.
  • Provided as a small-quantity vial for reference and method development.
Catalog No. 50-002-44644 Supplier Medchemexpress LLC Supplier No. HY10330S1MG
Toceranib-d8 is the deuterium-labeled isotopologue of toceranib, provided as a research-grade analytical standard for laboratory use. It is intended for use as a stable isotope internal standard in LC-MS/MS, pharmacokinetic, and tracer studies to enable accurate quantitation and differentiation from the unlabeled compound.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (>99%) suitable for analytical applications.
  • Intended for pharmacokinetic, bioanalytical, and tracer studies.
  • Provided as a small-quantity vial for reference and method development.