Learn More
Medchemexpress LLC Toceranib-d8 | 1795134-78-9 | 99.1% | 404.51 g/mol | C22H17D8FN4O2 | 5 MG
SDP
Catalog No. 5000244876
Encompass_Preferred
Toceranib-d8 is a deuterium-labeled stable isotope of toceranib supplied as a high-purity research standard for analytical and tracer applications. It is intended for research use only and is provided in small package sizes suitable for use as an internal standard in mass spectrometry and quantitative assays.
  • Deuterium-labeled stable isotope suitable for analytical use
  • High purity for reliable quantitation in MS assays
  • Available in small mass packages to minimize waste
  • Supplied with COA and datasheet for lot traceability
  • Intended for laboratory research use only
Catalog No. 50-002-44876 Supplier Medchemexpress LLC Supplier No. HY10330S5MG
Toceranib-d8 is a deuterium-labeled stable isotope of toceranib supplied as a high-purity research standard for analytical and tracer applications. It is intended for research use only and is provided in small package sizes suitable for use as an internal standard in mass spectrometry and quantitative assays.
  • Deuterium-labeled stable isotope suitable for analytical use
  • High purity for reliable quantitation in MS assays
  • Available in small mass packages to minimize waste
  • Supplied with COA and datasheet for lot traceability
  • Intended for laboratory research use only