Learn More
Selleck Chemical LLC Toceranib phosphate S5272-25mg
SDP
Catalog No. 503111726
Encompass_Preferred
Toceranib phosphate (Palladia SU11654) the phosphate salt of toceranib is a selective inhibitor of the tyrosine kinase activity of several members of the split kinase RTK family including Flk-1/KDR PDGFR and Kit with Ki values of 6 nM and 5 nM for Flk-1/KDR and PDGFR respectively
Catalog No. 50-311-1726 Supplier Selleck Chemical LLC Supplier No. S527225MG
Toceranib phosphate (Palladia SU11654) the phosphate salt of toceranib is a selective inhibitor of the tyrosine kinase activity of several members of the split kinase RTK family including Flk-1/KDR PDGFR and Kit with Ki values of 6 nM and 5 nM for Flk-1/KDR and PDGFR respectively