Learn More
Medchemexpress LLC HY-10330A 50mg Medchemexpress, Toceranib (phosphate) CAS:874819-74-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10330A50MG
Encompass_Preferred
Medchemexpress, HY-10330A 50mg Toceranib (phosphate) CAS:874819-74-6 Toceranib phosphate (SU11654 phosphate) is an orally active receptor tyrosine kinase (RTK) inhibitor, and it potently inhibits PDGFR, VEGFR, and Kit with Kis of 5 and 6 nM for PDGFRβ and Flk-1/KDR, respectively. Toceranib phosphate (SU11654 phosphate) has antitumor and antiangiogenic activity, and used in the treatment of canine mast cell tumors . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8071