Learn More
Medchemexpress LLC HY-P9917 1mg Medchemexpress, Tocilizumab CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99171MG
Encompass_Preferred
Medchemexpress, HY-P9917 1mg Tocilizumab CAS:0 Tocilizumab (Anti-Human IL6R, Humanized Antibody) is an anti-human interleukin-6 receptor (IL-6R) neutralizing antibody, prevents binding of IL-6 to the IL-6R, thereby inhibiting both classic and trans-signaling. Tocilizumab (Anti-Human IL6R, Humanized Antibody) can be used for the treatment of rheumatoid arthritis. Tocilizumab is remarkablely effective for the study of severe COVID-19 (coronavirus disease)[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9437