Learn More
Medchemexpress LLC HY-B0717 10g , Tocofersolan CAS:9002-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB071710G
Encompass_Preferred
HY-B0717 10g Tocofersolan CAS: 9002-96-4 Tocofersolan is synthetic polyethylene glycol derivative of α-tocopherol. Tocofersolan is an orally active and water-soluble analog of vitamin E. Tocofersolan can reduce neurobehavioral deficits in zebrafish embryos exposed to moderate and high concentrations of BaP during early development. Tocofersolan shows antioxidant activity. Tocofersolan can be used to provide an orally bioavailable source of vitamin E. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1143