Learn More
Medchemexpress LLC Tocofersolan | 9002-96-4 | 98.0% | 100 G
SDP
Catalog No. 5000408186
Encompass_Preferred
Tocofersolan is a synthetic polyethylene glycol derivative of α-tocopherol. It is an orally active and water-soluble analog of vitamin E. It has demonstrated the ability to reduce neurobehavioral deficits in zebrafish embryos when exposed to moderate and high concentrations of BaP during early development, exhibits antioxidant activity, and can serve as an orally bioavailable source of vitamin E.
  • Synthetic polyethylene glycol derivative of α-tocopherol
  • Orally active and water-soluble analog of vitamin E
  • Reduces neurobehavioral deficits in zebrafish embryos
  • Exhibits antioxidant activity
  • Serves as an orally bioavailable source of vitamin E
Catalog No. 50-004-08186 Supplier Medchemexpress LLC Supplier No. HYB0717100G
Tocofersolan is a synthetic polyethylene glycol derivative of α-tocopherol. It is an orally active and water-soluble analog of vitamin E. It has demonstrated the ability to reduce neurobehavioral deficits in zebrafish embryos when exposed to moderate and high concentrations of BaP during early development, exhibits antioxidant activity, and can serve as an orally bioavailable source of vitamin E.
  • Synthetic polyethylene glycol derivative of α-tocopherol
  • Orally active and water-soluble analog of vitamin E
  • Reduces neurobehavioral deficits in zebrafish embryos
  • Exhibits antioxidant activity
  • Serves as an orally bioavailable source of vitamin E