Learn More
Cayman Chemical Tocopherol 50mg
SDP
Supplier:  Cayman Chemical 2675850 MG
Encompass_Preferred
A lipid-soluble form of vitamin E with antioxidant activity; scavenges DPPH radicals (EC50 = 68.2 μM); inhibits LPS-induced COX-2 gene expression in mouse RAW 264.7 macrophage cells at 100 μM
Catalog No. 50-293-4947